missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl butyrate, 99%

Product Code. 11495827
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495827 250 mL 250mL
11485827 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495827 Supplier Thermo Scientific Alfa Aesar Supplier No. B24390.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-21-7 | C8H16O2 | 144.21 g/mol

It is very important for the food and beverages industries and used as perfuming agents. It is a applied as a solvent for resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is very important for the food and beverages industries and used as perfuming agents. It is a applied as a solvent for resins.

Solubility
Soluble in water. (1 g/L).

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-21-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00009450
InChI Key XUPYJHCZDLZNFP-UHFFFAOYSA-N
Synonym butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
PubChem CID 7983
ChEBI CHEBI:87429
IUPAC Name butyl butanoate
SMILES CCCCOC(=O)CCC

Specifications

Melting Point -92°C
Boiling Point 164°C to 165°C
Flash Point 49°C (120°F)
Linear Formula CH3CH2CH2CO2(CH2)3CH3
Quantity 250 mL
UN Number UN3272
Beilstein 1747101
Merck Index 14,1556
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material n-Butyl butyrate
Show More Show Less
compliance-icons
Product Identifier
  • n-Butyl butyrate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Butyl butyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.