missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Product Code. 11313588
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

n-Butyl acetate is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Color Colorless
Density 0.881 g/mL
pH 5
Boiling Point 124°C to 126°C
Flash Point 25°C (77°F)
Odor Fruit-like
Linear Formula CH3CO2(CH2)3CH3
Refractive Index 1.3945
Quantity 250 mL
UN Number UN1123
Beilstein 1741921
Merck Index 14,1535
Solubility Information Miscible with ethanol,ethyl ether,alcohol,acetone,chloroform and most hydrocarbonsSlightly miscible with water.
Formula Weight 116.16
Percent Purity ≥99.5%
Grade HPLC
Physical Form Liquid
Chemical Name or Material n-Butyl acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'