missing translation for 'onlineSavingsMsg'
Learn More

N-Butyl Acetate, 99+%, for Electronic Use, Fisher Chemical™

Product Code. 11493543 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
5 L
Packaging:
Aluminum can
Amber glass bottle
Unit Size:
2.5 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11493543 2.5 L Amber glass bottle 2.5 L
10699263 5 L Aluminum can 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493543 Supplier Fisher Chemical Supplier No. B/4960/17
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 123-86-4
MDL Number 9445

Specifications

Melting Point -90°C
Color Colorless
Boiling Point 126°C
Packaging Amber glass bottle
Molecular Formula C6H12O2
Quantity 2.5 L
Formula Weight 116.16g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • n-Butyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.