missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl 4-aminobenzoate, 99%

Product Code. 15450908
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 15450908

Brand: Thermo Scientific Alfa Aesar H54690.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-25-7 | C11H15NO2 | 193.246 g/mol

n-Butyl 4-aminobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl 4-aminobenzoate is used as pharmaceutical intermediate.

Solubility
Very slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 94-25-7
Melting Point 56°C to 59°C
Boiling Point 174°C (8 mmHg)
Molecular Formula C11H15NO2
MDL Number MFCD00017112
Quantity 100 g
Beilstein 1211465
Sensitivity Light sensitive
Merck Index 14,1512
Synonym butamben, butesin, butylcaine, butyl aminobenzoate, butesine, butoform, planoform, scuroform, scuroforme, butyl keloform
Solubility Information Very slightly soluble in water.
InChI Key IUWVALYLNVXWKX-UHFFFAOYSA-N
SMILES CCCCOC(=O)C1=CC=C(C=C1)N
IUPAC Name butyl 4-aminobenzoate
Molecular Weight (g/mol) 193.246
PubChem CID 2482
ChEBI CHEBI:3231
Formula Weight 193.25
Percent Purity 99%
Chemical Name or Material n-Butyl 4-aminobenzoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.