missing translation for 'onlineSavingsMsg'
Learn More

n-Butanol (Clear, Colorless Liq.), Fisher BioReagents

Product Code. 10420074
Change view
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
25mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420074 25 mL Glass Bottle 25mL
10294763 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420074 Supplier Fisher BioReagents Supplier No. BP50525
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
n-Butanol is used as an analytical reagent and solvent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -89°C
Color Colorless
Acidity 0.005% max.
Density 0.810g/cm³
Quantity 25 mL
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥99.8 %
Linear Formula CH3CH2CH2CH2OH
Vapor Pressure 6.7mbar at 20°C
Viscosity 2.95 mPa/s at 20°C
UN Number 1120
Water 0.1% max.
Merck Index 15, 1542
Boiling Point 117.6°C
Formula Weight 74.12
Percent Purity ≥99.8%
Chemical Name or Material n-Butanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.