missing translation for 'onlineSavingsMsg'
Learn More

n-Butanol, 99% max., Molecular biology reagent grade, MP Biomedicals™

Product Code. 11406461 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
Unit Size:
1 L
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406461 25 mL 25mL
11416461 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406461 Supplier MP Biomedicals Supplier No. 0219400125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Density 0.81g/cm3 at 21°C
Quantity 25 mL
Physical Form Liquid
Formula Weight 74.12
Percent Purity 99% max.
Grade Molecular Biology
Chemical Name or Material n-Butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.