missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosuccinimide, TRC

Product Code. 30006018
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006018 100 g 100g
30007295 5 g 5g
30007121 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006018 Supplier Toronto Research Chemicals Supplier No. TRCB686995100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-08-5
Molecular Formula C4H4BrNO2
Molecular Weight (g/mol) 177.98
Synonym 1-Bromo-2,5-pyrrolidinedione, Bromosuccinimide, NBS, NSC 16, Succinbromimide;
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES BrN1C(=O)CCC1=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H4BrNO2/c5-6-3(7)1-2-4(6)8/h1-2H2
Formula Weight 176.9425
Chemical Name or Material N-Bromosuccinimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.