missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosuccinimide, 99%

Product Code. 11478456
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478456 250 g 250g
11468456 1000 g 1000g
11488456 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478456 Supplier Thermo Scientific Alfa Aesar Supplier No. A15922.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128-08-5 | C4H4BrNO2 | 177.985 g/mol

N-Bromosuccinimide is a chemical reagent used in electrophilic additions and radical substitution reactions in synthetic organic chemistry. In Wohl-Ziegler reaction, it is involved in allylic and benzylic bromination reaction. It is also used in the alfa-bromination of carbonyl derivatives. In the Hofmann rearrangement, it is used in the presence of a strong base, such as 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) reacts with primary amides to produce a carbamate. It oxidizes silyl ethers to aldehydes in the presence of azobisisobutyronitrile. It is also used for the modification of ribosomal sulfhydryl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Bromosuccinimide is a chemical reagent used in electrophilic additions and radical substitution reactions in synthetic organic chemistry. In Wohl-Ziegler reaction, it is involved in allylic and benzylic bromination reaction. It is also used in the alfa-bromination of carbonyl derivatives. In the Hofmann rearrangement, it is used in the presence of a strong base, such as 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) reacts with primary amides to produce a carbamate. It oxidizes silyl ethers to aldehydes in the presence of azobisisobutyronitrile. It is also used for the modification of ribosomal sulfhydryl groups.

Solubility
Soluble in acetone, tetrahydrofuran, dimethyl formamide, dimethyl sulfoxide and acetonitrile. Slightly soluble in water and acetic acid. Insoluble in ether, hexane and carbon tetrachloride.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, iron, iron salts, halogenated hydrocarbons, amines and metals. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-08-5
Molecular Formula C4H4BrNO2
Molecular Weight (g/mol) 177.985
MDL Number MFCD00005510
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
Synonym n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide
PubChem CID 67184
ChEBI CHEBI:53174
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)Br

Specifications

Melting Point 175°C to 180°C (decomposition)
Density 2.098
Assay Percent Range 99%
Quantity 250 g
UN Number UN3261
Beilstein 113916
Sensitivity Light sensitive
Merck Index 14,1438
Solubility Information Soluble in acetone,tetrahydrofuran,dimethyl formamide,dimethyl sulfoxide and acetonitrile. Slightly soluble in water and acetic acid. Insoluble in ether,hexane and carbon tetrachloride.
Formula Weight 177.99
Percent Purity 99%
Chemical Name or Material N-Bromosuccinimide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Bromosuccinimide
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contain: N-Bromosuccinimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.