missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosuccinimide, 99%

Product Code. 10132900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
10g
1kg
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132900 1 kg Glass bottle 1kg
10559820 10 g Glass bottle 10g
10655332 100 g Glass bottle 100g
10478820 500 g Glass bottle 500g
11925581 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10132900 Supplier Thermo Scientific Acros Supplier No. 107450010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 128-08-5 | C4H4BrNO2 | 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent, C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-08-5
MDL Number MFCD00005510
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
Synonym n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide
PubChem CID 67184
ChEBI CHEBI:53174
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 175.0°C to 178.0°C
Color White to Yellow
Flash Point 180°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (on dry substance) (Iodometry)
Packaging Glass bottle
Molecular Formula C4H4BrNO2
Quantity 1 kg
Beilstein 21, 380
Fieser 01,78; 02,40; 03,34; 04,49; 05,65; 06,74; 07,37; 08,54; 09,70; 10,57; 11,79; 12,79; 13,41; 14,57; 15,50; 16,49
Merck Index 15, 1448
Solubility Information Solubility in water: 14.8g/L (20°C). Other solubilities: 3.10g/100g glacial acetic acid(25°C),g/100g solvent (25°C):,tert. butanol:0.73- aceton:14.40,carbone tetrachloride:0.02- hexane:0.006,benzene:0.5
Molecular Weight (g/mol) 177.99
Formula Weight 177.99
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Bromosuccinimide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Bromosuccinimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.