missing translation for 'onlineSavingsMsg'
Learn More

N-Bromophthalimide, 98+%

Product Code. 11474683
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474683 100 g 100g
11454683 5 g 5g
11464683 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474683 Supplier Thermo Scientific Alfa Aesar Supplier No. A11601.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2439-85-2 | C8H4BrNO2 | 226.029 g/mol

Brominating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2439-85-2
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00005888
InChI Key MARXMDRWROUXMD-UHFFFAOYSA-N
PubChem CID 75542
IUPAC Name 2-bromoisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)Br

Specifications

Melting Point 204°C to 208°C
Quantity 100 g
Beilstein 131211
Sensitivity Light and moisture sensitive
Synonym n-bromophthalimide, 2-bromoisoindoline-1,3-dione, 2-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-bromo, n-bromophtalimide, n-bromo phthalimide, acmc-1cjlt, 2-bromobenzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,2-bromo
Formula Weight 226.03
Percent Purity ≥98%
Chemical Name or Material N-Bromophthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.