missing translation for 'onlineSavingsMsg'
Learn More

N-(Bromomethyl)phthalimide, 95%

Product Code. 11377855
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377855 50 g 50g
11367855 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377855 Supplier Thermo Scientific Alfa Aesar Supplier No. L02818.18
  • ONLINE EXCLUSIVE PRICE
    £143.00
    £128.65 / 50g
    Save £14.35
    10% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5332-26-3 | C9H6BrNO2 | 240.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5332-26-3
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD00005897
InChI Key UUSLLECLCKTJQF-UHFFFAOYSA-N
Synonym n-bromomethyl phthalimide, 2-bromomethyl isoindoline-1,3-dione, n-bromomethylphthalimide, phthalimide, n-bromomethyl, phthalimidomethyl bromide, 1h-isoindole-1,3 2h-dione, 2-bromomethyl, 2-bromomethyl isoindole-1,3-dione, 2-bromomethyl-1h-isoindole-1,3 2h-dione, 2-bromomethyl-2,3-dihydro-1h-isoindole-1,3-dione, acmc-1awzk
PubChem CID 79244
IUPAC Name 2-(bromomethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CBr

Specifications

Melting Point 144°C to 150°C
Odor Odorless
Quantity 50 g
Beilstein 140943
Sensitivity Light and moisture sensitive
Formula Weight 240.07
Percent Purity 95%
Chemical Name or Material N-(Bromomethyl)phthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.