missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-bromoacetamide, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N-bromoacetamide |
| CAS | 79-15-2 |
| Molecular Formula | C2H4BrNO |
| Quantity | 5 g |
| Synonym | Acetobromamide, NBA |
| InChI Formula | InChI=1S/C2H4BrNO/c1-2(5)4-3/h1H3,(H,4,5) |
| SMILES | CC(=O)NBr |
| IUPAC Name | N-bromoacetamide |
| Molecular Weight (g/mol) | 137.96 |
| Formula Weight | 136.9476 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction