missing translation for 'onlineSavingsMsg'
Learn More

N-Bromoacetamide, 98%, Thermo Scientific™

Product Code. 11945701 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 11945701

Brand: Thermo Scientific Acros 159050500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 79-15-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 102°C to 105°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C2H4BrNO
Linear Formula CH3CONHBr
Quantity 50 g
Fieser 01,74; 02,39; 04,47; 05,61
Synonym acetamide, n-bromo, unii-0aq6mwh7zb, ccris 4590, 0aq6mwh7zb, acetobromamide, n-bromo-acetamide, sjyhiabiktp@, acmc-209pgg, n-bromoacetamide, powder, lopac-b-2377
InChI Key VBTQNRFWXBXZQR-UHFFFAOYSA-N
SMILES CC(=O)NBr
IUPAC Name N-bromoacetamide
Molecular Weight (g/mol) 137.964
PubChem CID 4353
Formula Weight 137.96
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material N-Bromoacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Bromoacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.