missing translation for 'onlineSavingsMsg'
Learn More

N-Bromoacetamide, 95%

Product Code. 11397905
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11397905

Brand: Thermo Scientific Alfa Aesar L02953.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-15-2 | C2H4BrNO | 137.964 g/mol

Abolishes the rapid inactivation of membrane sodium- and potassium-ion channels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Abolishes the rapid inactivation of membrane sodium- and potassium-ion channels.

Solubility
Soluble in water.

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-15-2
Molecular Formula C2H4BrNO
Molecular Weight (g/mol) 137.964
MDL Number MFCD00037097
InChI Key VBTQNRFWXBXZQR-UHFFFAOYSA-N
Synonym acetamide, n-bromo, unii-0aq6mwh7zb, ccris 4590, 0aq6mwh7zb, acetobromamide, n-bromo-acetamide, sjyhiabiktp@, acmc-209pgg, n-bromoacetamide, powder, lopac-b-2377
PubChem CID 4353
IUPAC Name N-bromoacetamide
SMILES CC(=O)NBr

Specifications

Melting Point 102°C to 106°C
Linear Formula CH3CONHBr
Quantity 5 g
Beilstein 969346
Merck Index 14,1398
Solubility Information Soluble in water.
Formula Weight 137.97
Percent Purity 95%
Chemical Name or Material N-Bromoacetamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.