missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-trans-4-N-FMOC-amino-L-proline, 97%

Product Code. 10546864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10546864 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10546864 Supplier Thermo Scientific Acros Supplier No. 426040010
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 176486-63-8 | C25H28N2O6 | 452.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 176486-63-8
CAS Min % 96.0
CAS Max % 100.0
Melting Point 199°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C25H28N2O6
MDL Number MFCD00673783
Quantity 1 g
Specific Rotation -33° to -35° (20°C, 589nm) (c=1, chloroform)
Synonym 2s,4r-4-9h-fluoren-9-yl methoxy carbonyl amino-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-boc-trans-4-n-fmoc-amino-l-proline, 2s,4r-n-tert-butoxycarbonyl-4-n-9-fluorenylmethoxycarbonyl aminopyrrolidine-2-carboxylic acid, 2s,4r-4-9h-fluoren-9-yl methoxy carbonylamino-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, 2s,4r-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino pyrrolidine-2-carboxylic acid, ambotzbaa1783, 2s,4r-fmoc-4-amino-1-boc-pyrrolidine-2-carboxyli, 2s,4r-1-boc-4-n-fmoc-4-aminopyrrolidine-2-carboxylic acid, 2s,4r-trans-n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-proline
InChI Key UPXRTVAIJMUAQR-VFNWGFHPSA-N
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
IUPAC Name (2S,4R)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 452.5
PubChem CID 2756135
Formula Weight 452.5
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-trans-4-N-FMOC-amino-L-proline
Show More Show Less
Product Identifier
  • N-BOC-trans-4-N-FMOC-amino-L-proline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.