missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-trans-4-fluoro-L-proline methyl ester, 97%, Thermo Scientific™

Product Code. 15417725
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417725 250 mg 250mg
11352899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417725 Supplier Thermo Scientific Alfa Aesar Supplier No. H27747.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-trans-4-fluoro-L-proline methyl ester is used to prepare (S)-2-[N-(N′-benzyl-(S)-4-fluoro-L-prolyl)amino]benzophenone by reacting with of 2-aminobenzophenone in the presence of potassium tert -butoxide.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203866-18-6
Molecular Formula C11H18FNO4
Molecular Weight (g/mol) 247.266
MDL Number MFCD07367993
InChI Key METPQQHVRNLTRX-SFYZADRCSA-N
Synonym 2s,4r-1-tert-butyl 2-methyl 4-fluoropyrrolidine-1,2-dicarboxylate, n-boc-trans-4-fluoro-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-fluoropyrrolidine-1,2-dicarboxylate, 4r-1-boc-4-fluoro-l-proline methyl ester, methyl n-tert-butoxycarbonyl-2s, 4r-4-fluoroprolinate, 1-tert-butyl 2-methyl 2s,4r-4-fluoro-1,2-pyrrolidinedicarboxylate, boc-trans-4-fluoro-l-proline methyl ester, n-t-boc-trans-4-fluoro-l-proline methyl ester, 1-tert-butoxycarbonyl-4beta-fluoro-l-proline methyl ester
PubChem CID 11806976
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4R)-4-fluoropyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)F

Specifications

Melting Point 36°C to 39°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 247.27
Percent Purity 97%
Chemical Name or Material N-Boc-trans-4-fluoro-L-proline methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.