missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-trityl-D-cysteine, 98%

Product Code. p-7041624
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15429318

Brand: Thermo Scientific Alfa Aesar H62863.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87494-13-1 | C27H29NO4S | 463.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87494-13-1
Molecular Formula C27H29NO4S
Molecular Weight (g/mol) 463.59
MDL Number MFCD00236839
InChI Key JDTOWOURWBDELG-UHFFFAOYNA-N
Synonym boc-d-cys trt-oh, boc-s-trityl-d-cysteine, n-alpha-t-butyloxycarbonyl-s-trityl-d-cysteine, s-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-l-cysteine trityl, 2s-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, boc-cys trt, boc-cys-trt, ambotzbaa5000, n-boc-s-trityl-d-cysteine
PubChem CID 11590774
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Quantity 1 g
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
Formula Weight 463.59
Percent Purity 98%
Chemical Name or Material N-Boc-S-trityl-D-cysteine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.