missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-benzyl-L-cysteine, 98%, Thermo Scientific Chemicals

Product Code. 15470579
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5068-28-0
Molecular Formula C15H21NO4S
Molecular Weight (g/mol) 311.40
MDL Number MFCD00065567
InChI Key IFVORPLRHYROAA-UHFFFAOYNA-N
Synonym boc-s-benzyl-l-cysteine, boc-cys bzl-oh, r-3-benzylthio-2-tert-butoxycarbonyl amino propanoic acid, n-boc-s-benzyl-l-cysteine, n-tert-butoxycarbonyl-s-benzyl-l-cysteine, s-benzyl-n-tert-butoxycarbonyl-l-cysteine, 2r-3-benzylsulfanyl-2-tert-butoxycarbonyl amino propanoic acid, boc-s-benzyl-d-cys, boc-s-benzyl-l-cys, alanine, 3-benzylthio-n-carboxy-, n-tert-butyl ester, l
PubChem CID 2724757
SMILES CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 86°C to 88°C
Beilstein 2000417
IUPAC Name (2R)-3-benzylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Formula Weight 311.4
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Boc-S-benzyl-L-cysteine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'