missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-pyrrolidine, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | N-Boc-pyrrolidine |
| CAS | 86953-79-9 |
| Molecular Formula | C9H17NO2 |
| Quantity | 25 g |
| Synonym | 1-Pyrrolidinecarboxylic Acid 1,1-Dimethylethyl Ester, 1-(tert-Butoxycarbonyl)pyrrolidine, N-tert-Butoxycarbonylpyrrolidine, NT 0186, tert-Butyl 1-Pyrrolidinecarboxylate |
| InChI Formula | InChI=1S/C9H17NO2/c1-9(2,3)12-8(11)10-6-4-5-7-10/h4-7H2,1-3H3 |
| SMILES | CC(C)(C)OC(=O)N1CCCC1 |
| IUPAC Name | tert-butyl pyrrolidine-1-carboxylate |
| Molecular Weight (g/mol) | 171.24 |
| Formula Weight | 171.1259 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction