missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Pyrrole, 98%

Product Code. 11996661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11996661 25 g 25g
11906671 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11996661 Supplier Thermo Scientific Acros Supplier No. 438510250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5176-27-2 | C9H13NO2 | 167.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5176-27-2
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
InChI Key IZPYBIJFRFWRPR-UHFFFAOYSA-N
Synonym tert-butyl 1h-pyrrole-1-carboxylate, n-boc-pyrrole, n-t-boc-pyrrole, tert-butyl 1-pyrrolecarboxylate, t-butyl 1h-pyrrole-1-carboxylate, 1h-pyrrole-1-carboxylic acid, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl-1h-pyrrole, 1-boc pyrrole, 1-boc-pyrrole, acmc-209kvp
PubChem CID 643494
IUPAC Name tert-butyl pyrrole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=CC=C1

Specifications

Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 97.0°C (2.3 Torr)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1
Formula Weight 167.21
Percent Purity 98%
Physical Form Solution
Chemical Name or Material N-BOC-Pyrrole
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-Pyrrole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.