missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-tert-butyl-L-threonine, 95%

Product Code. 15469198
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15469198

Brand: Thermo Scientific Alfa Aesar H62713.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13734-40-2 | C13H25NO5 | 275.35 g/mol

N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13734-40-2
Molecular Formula C13H25NO5
Molecular Weight (g/mol) 275.35
MDL Number MFCD00151115
InChI Key LKRXXARJBFBMCE-BDAKNGLRSA-N
Synonym boc-thr tbu-oh, boc-o-tert-butyl-l-threonine, boc-o-t-butyl-l-threonine, 2s,3r-3-tert-butoxy-2-tert-butoxycarbonyl amino butanoic acid, l-threonine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, boc-thr t-bu-oh, pubchem12289, boc-l-thr tbu-oh, boc-o-tert-butyl-l-thr-oh
PubChem CID 7015770
IUPAC Name (2S,3R)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 95°C to 98°C
Beilstein 4454820
Solubility Information Soluble in DMF.
Quantity 5 g
Formula Weight 275.34
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-O-tert-butyl-L-threonine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.