missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-methyl-L-tyrosine, 98%

Product Code. p-7053992
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15498398 1 g 1g
15408408 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15498398

Brand: Thermo Scientific Alfa Aesar H61820.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53267-93-9 | C15H21NO5 | 295.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53267-93-9
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.34
MDL Number MFCD00065603
InChI Key SLWWWZWJISHVOU-STGVRZAANA-N
Synonym boc-tyr me-oh, boc-o-methyl-l-tyrosine, 2s-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid, boc-4-methoxyphenylalanine, s-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid, boc-l-4-methoxyphenylalanine, boc-4-methoxy-phe-oh, boc-4-methoxy-l-phenylalanine, n-tert-butoxycarbonyl-o-methyltyrosine, n-alpha-t-butyloxycarbonyl-o-methyl-l-tyrosine
PubChem CID 2762280
IUPAC Name (2S)-3-(4-methoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES COC1=CC=C(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)C=C1

Specifications

Quantity 1 g
Formula Weight 295.33
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-O-methyl-L-tyrosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.