missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-ethyl-L-serine, 97%

Product Code. 15499517
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499517 250 mg 250mg
15489517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499517 Supplier Thermo Scientific Alfa Aesar Supplier No. H52781.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104839-00-1 | C10H19NO5 | 233.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104839-00-1
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.264
MDL Number MFCD06797558
InChI Key KNBDVSSESAKBJM-ZETCQYMHSA-N
Synonym boc-s-2-amino-3-ethoxypropionic acid, s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, 2s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, boco-etser, serine, n-1,1-dimethylethoxy carbonyl-o-ethyl, n-tert-butoxycarbonyl-o-ethylserine, n-boc-2-amino-3-ethoxy-propionic acid, s-n-boc-2-amino-3-ethoxypropionic acid, s-2-tertbutoxycarbonylamino-3-ethoxypropionic acid, n-1,1-dimethylethyl oxy carbonyl-o-ethylserine
PubChem CID 15058451
IUPAC Name (2S)-3-ethoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CCOCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N-Boc-O-ethyl-L-serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.