missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-α-BOC-N-omega-Tosyl-L-arginine, 97%, contains <15% ethyl acetate, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 99°C to 100°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Quantity | 5 g |
| Specific Rotation | -3.2 |
| Residual Solvents | 15.0% max. (EtOAc) |
| Formula Weight | 428.51 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction