missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-BOC-N-Methyl-L-phenylalanine, TRC
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.50g
Specifications
Specifications
| Chemical Name or Material | N-BOC-N-Methyl-L-phenylalanine |
| CAS | 37553-65-4 |
| Molecular Formula | C15H21NO4 |
| Quantity | 2.5 g |
| Synonym | N-(tert-Butoxycarbonyl)-N-methylphenylalanine, N-(tert-Butoxycarbonyl)-N-methyl-L-phenylalanine, N-[(1,1-Dimethylethoxy)carbonyl]-N-methyl-L-phenylalanine, N-Methyl-N-(tert-butoxycarbonyl)-L-phenylalanine |
| InChI Formula | InChI=1S/C15H21NO4/c1-15(2,3)20-14(19)16(4)12(13(17)18)10-11-8-6-5-7-9-11/h5-9,12H,10H2,1-4H3,(H,17,18)/t12-/m0/s1 |
| SMILES | CN([C@@H](Cc1ccccc1)C(=O)O)C(=O)OC(C)(C)C |
| IUPAC Name | (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-phenylpropanoic acid |
| Molecular Weight (g/mol) | 279.33 |
| Formula Weight | 279.1471 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction