missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-L-phenylalanine, 95%

Product Code. p-7054203
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15498768

Brand: Thermo Scientific Alfa Aesar H62221.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37553-65-4 | C15H21NO4 | 279.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37553-65-4
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD00038754
InChI Key AJGJINVEYVTDNH-LBPRGKRZSA-N
Synonym boc-n-me-phe-oh, boc-n-methyl-l-phenylalanine, boc-mephe-oh, boc-l-mephe-oh, phenylalanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, bocmepheoh, ambotzbaa1113, boc me phe oh, pubchem12252
PubChem CID 7010602
IUPAC Name (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)N(C)C(CC1=CC=CC=C1)C(=O)O

Specifications

Beilstein 2386503
Quantity 1 g
Formula Weight 279.33
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-phenylalanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.