missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-1,3-diaminopropane, 95%, Thermo Scientific Chemicals

Product Code. 15462839
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462839 1 g 1g
15472839 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462839 Supplier Thermo Scientific Alfa Aesar Supplier No. H66932.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 150349-36-3 | Molecular Formula: C9H20N2O2 | Molecular Weight: 188.271 g/mol

It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150349-36-3
Molecular Formula C9H20N2O2
Molecular Weight (g/mol) 188.271
MDL Number MFCD01631189
InChI Key PNQYAMWGTGWJDW-UHFFFAOYSA-N
Synonym n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
PubChem CID 2756543
IUPAC Name tert-butyl N-(3-aminopropyl)-N-methylcarbamate
SMILES CC(C)(C)OC(=O)N(C)CCCN

Specifications

Color Yellow
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 188.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Boc-N-methyl-1,3-diaminopropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.