missing translation for 'onlineSavingsMsg'
Learn More

N-Boc Linagliptin, TRC

Product Code. 30137198
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137198 50 mg 50mg
30134931 25 mg 25mg
30010693 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137198 Supplier Toronto Research Chemicals Supplier No. TRCB65681050MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 668273-75-4
Molecular Formula C30 H36 N8 O4
Molecular Weight (g/mol) 572.66
Synonym N-[(3R)-1-[7-(2-Butyn-1-yl)-2,3,6,7-tetrahydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-2,6-dioxo-1H-purin-8-yl]-3-piperidinyl]carbamic Acid 1,1-Dimethylethyl Ester
IUPAC Name tert-butyl N-[(3R)-1-[7-but-2-ynyl-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]-2,6-dioxopurin-8-yl]piperidin-3-yl]carbamate
SMILES CC#CCn1c(nc2N(C)C(=O)N(Cc3nc(C)c4ccccc4n3)C(=O)c12)N5CCC[C@H](C5)NC(=O)OC(C)(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C30H36N8O4/c1-7-8-16-37-24-25(34-27(37)36-15-11-12-20(17-36)32-28(40)42-30(3,4)5)35(6)29(41)38(26(24)39)18-23-31-19(2)21-13-9-10-14-22(21)33-23/h9-10,13-14,20H,11-12,15-18H2,1-6H3,(H,32,40)/t20-/m1/s1
Formula Weight 572.286
Percent Purity >95%
Chemical Name or Material N-Boc Linagliptin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.