missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-serine methyl ester, 95%

Product Code. 15466328
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466328 10 g 10g
15456328 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466328 Supplier Thermo Scientific Alfa Aesar Supplier No. H59889.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2766-43-0 | C9H17NO5 | 219.237 g/mol

N-tert-Butoxycarbonyl-L-serine Methyl Ester is an amino acid derivative used in the preparation of Tn antigen, L-glutamic acid p-nitroanilide analogs and new biomedical polymers having Serine and Threonine side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-tert-Butoxycarbonyl-L-serine Methyl Ester is an amino acid derivative used in the preparation of Tn antigen, L-glutamic acid p-nitroanilide analogs and new biomedical polymers having Serine and Threonine side groups.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2766-43-0
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00191869
InChI Key SANNKFASHWONFD-LURJTMIESA-N
Synonym boc-ser-ome, boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, n-boc-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-l-ser-ome, methyl tert-butoxycarbonyl-l-serinate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-ser-ome; boc-l-serine methyl ester
PubChem CID 7019177
IUPAC Name methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC

Specifications

Density 1.082
Flash Point 113°C (235°F)
Linear Formula HOCH2CH[NHCO2C(CH3)3]CO2CH3
Refractive Index 1.4525
Quantity 10 g
Beilstein 3545389
Solubility Information Slightly soluble in water.
Optical Rotation −18° (c=5 in Methanol)
Formula Weight 219.23
Percent Purity 95%
Chemical Name or Material N-Boc-L-serine methyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.