missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-prolinol, 98+%

Product Code. 11392327
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392327 1 g 1g
15408919 5 g 5g
15418919 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392327 Supplier Thermo Scientific Alfa Aesar Supplier No. L09885.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69610-40-8 | C10H19NO3 | 201.266 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69610-40-8
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD00066232
InChI Key BFFLLBPMZCIGRM-QMMMGPOBSA-N
Synonym n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
PubChem CID 643448
IUPAC Name tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO

Specifications

Melting Point 59°C to 62°C
Quantity 1 g
Beilstein 3542667
Optical Rotation −54° (c=5 in Methanol)
Formula Weight 201.27
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-prolinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.