missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-proline, TRC

Product Code. 30332872
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332872 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332872 Supplier Toronto Research Chemicals Supplier No. TRCB66420050G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-L-proline
CAS 15761-39-4
Molecular Formula C10H17NO4
Quantity 50 g
Synonym (2S)-1,2-Pyrrolidinedicarboxylic acid, 1-(1,1-dimethylethyl) ester, (S)-1,2-Pyrrolidinedicarboxylic acid, 1-(1,1-dimethylethyl) ester, L-1,2-Pyrrolidinedicarboxylic acid, 1-tert-butyl ester, (2S)-1,2-Pyrrolidinedicarboxylic acid 1-(1,1-dimethylethyl) ester, (2S)-1-Boc-pyrrolidine-2-carboxylic acid, (2S)-1-tert-Butoxycarbonylpyrrolidine-2-carboxylic acid, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinecarboxylic acid, (S)-1-(tertbutoxycarbonyl)pyrrolidine-2-carboxylic acid, (S)-1-Boc-beta-proline, (S)-N-tert-Butoxycarbonylproline, (S)-Pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 1-(tert-Butoxycarbonyl)-(S)-proline, 1-tert-Butoxycarbonyl-L-proline, 1-tert-Butyl (S)-pyrrolidine-1,2-dicarboxylate, 1-tert-Butyloxycarbonyl-L-proline, BOC-L-Proline, Boc-L-Pro-OH, L-N-Boc-proline, N-(t-Butoxycarbonyl)proline, N-(tert-Butoxycarbonyl)-(S)-proline, N-(tert-Butoxycarbonyl)-L-proline, N-(tert-Butoxycarbonyl)proline, N-(tert-Butyloxycarbonyl)-L-proline, N-Boc-(S)-proline, N-[(1,1-Dimethylethoxy)carbonyl]-(S)-proline, NSC 164660
InChI Formula InChI=1S/C10H17NO4/c1-10(2,3)15-9(14)11-6-4-5-7(11)8(12)13/h7H,4-6H2,1-3H3,(H,12,13)/t7-/m0/s1
SMILES CC(C)(C)OC(=O)N1CCC[C@H]1C(=O)O
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 215.25
Formula Weight 215.1158
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.