missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-L-Phenylalaninal, 97%, Thermo Scientific™

Product Code. 10306324 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 10306324

Brand: Thermo Scientific Acros 376742500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 86.0°C to 89.0°C
CAS 72155-45-4
CAS Min % 96.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C14H19NO3
Packaging Glass Bottle
MDL Number MFCD00143854
Specific Rotation − 45.00
Synonym n-boc-l-phenylalaninal, boc-phe-cho, boc-l-phenylalaninal, n-boc-phenylalaninal, boc-phenylalaninal, s-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, chembl94327, s---2-tert-butoxycarbonylamino-3-phenylpropanal, s-tert-butyl 1-oxo-3-phenylpropan-2-ylcarbamate, tert-butyl n-2s-1-oxo-3-phenylpropan-2-yl carbamate
InChI Key ZJTYRNPLVNMVPQ-LBPRGKRZSA-N
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C=O
IUPAC Name tert-butyl N-[(2S)-1-oxo-3-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol) 249.31
Quantity 250 mg
PubChem CID 6998013
Formula Weight 249.31
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-L-Phenylalaninal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.