missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-leucinol, 97%

Product Code. 11344276
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344276 1 g 1g
11354276 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344276 Supplier Thermo Scientific Alfa Aesar Supplier No. L19352.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82010-31-9 | C11H23NO3 | 217.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82010-31-9
Molecular Formula C11H23NO3
Molecular Weight (g/mol) 217.31
MDL Number MFCD00076950
InChI Key LQTMEOSBXTVYRM-VIFPVBQESA-N
Synonym boc-l-leucinol, n-boc-l-leucinol, boc-leucinol, boc-leu-ol, tert-butyl n-2s-1-hydroxy-4-methylpentan-2-yl carbamate, n-boc l-leucinol, pubchem12195, l-leucinol, n-boc protected, n-t-butoxycarbonyl-l-leucinol
PubChem CID 7018766
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
SMILES CC(C)C[C@@H](CO)NC(=O)OC(C)(C)C

Specifications

Boiling Point 212°C to 214°C
Refractive Index 1.45
Beilstein 4178460
Density 0.98
Quantity 1 g
Flash Point >110°C (230°F)
Formula Weight 217.31
Optical Rotation −27.5° (c=2 in Methanol)
Percent Purity 97%
Chemical Name or Material N-Boc-L-leucinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.