missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-glutamic Acid 1,5-Dimethyl Ester, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N-Boc-L-glutamic Acid 1,5-Dimethyl Ester |
| CAS | 59279-60-6 |
| Molecular Formula | C12H21NO6 |
| Quantity | 5 g |
| Synonym | Boc-Glu(OMe)-OMe, Dimethyl (2S)-2-(tert-Butoxycarbonylamino)pentanedioate, Dimethyl N-tert-Butoxycarbonyl-L-glutamate, Methyl (2S)-2-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoate, N-tert-Butoxycarbonyl-L-glutamic Acid Dimethyl Ester |
| InChI Formula | InChI=1S/C12H21NO6/c1-12(2,3)19-11(16)13-8(10(15)18-5)6-7-9(14)17-4/h8H,6-7H2,1-5H3,(H,13,16)/t8-/m0/s1 |
| SMILES | COC(=O)CC[C@H](NC(=O)OC(C)(C)C)C(=O)OC |
| IUPAC Name | dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate |
| Molecular Weight (g/mol) | 275.3 |
| Formula Weight | 275.1369 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction