missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid 1,5-dimethyl ester, 97%

Product Code. 15478638
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15478638

Brand: Thermo Scientific Alfa Aesar H62056.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59279-60-6 | C12H21NO6 | 275.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59279-60-6
Molecular Formula C12H21NO6
Molecular Weight (g/mol) 275.301
MDL Number MFCD03094774
InChI Key QNSPKWUAZQIIGZ-QMMMGPOBSA-N
Synonym boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester
PubChem CID 2734644
IUPAC Name dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC

Specifications

Color Colorless to Yellow
Quantity 5 g
Formula Weight 275.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Boc-L-glutamic acid 1,5-dimethyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.