missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-aspartic acid 4-tert-butyl 1-(N-succinimidyl) ester, 95%, Thermo Scientific™

Product Code. 15417106
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50715-50-9
Molecular Formula C17H26N2O8
Molecular Weight (g/mol) 386.401
MDL Number MFCD00063389
InChI Key PIITZDSTZQZNQH-JTQLQIEISA-N
Synonym boc-asp otbu-osu, l-aspartic acid,n-1,1-dimethylethoxy carbonyl-, 4-1,1-dimethylethyl 1-2,5-dioxo-1-pyrrolidinyl ester, 1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3s-3-tert-butoxycarbonyl amino butanedioate, boc-asp otbu-o-nhs, boc-asp otbu-osu c/n, boc-l-aspartic acida-n-hydroxysuccinimide ester, boc-l-aspartic acid 4-tert-butyl 1-hydroxysuccinimide ester, n-alpha-t-butyloxycarbonyl-l-aspartic acid alpha-succinimidyl beta-t-butyl ester
PubChem CID 13178396
IUPAC Name 4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioate
SMILES CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 98°C to 102°C
Color White
Beilstein 5361818
Formula Weight 386.4
Percent Purity 95%
Quantity 1 g
Sensitivity Moisture sensitive
Physical Form Powder
Chemical Name or Material N-Boc-L-aspartic acid 4-tert-butyl 1-(N-succinimidyl) ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'