missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-L-Asparagine 4-nitrophenyl ester, 98%, Thermo Scientific™

Product Code. 11925511 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11925511 1 g 1g
11935511 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11925511 Supplier Thermo Scientific Acros Supplier No. 440850010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4587-33-1
Molecular Formula C15H19N3O7
Molecular Weight (g/mol) 353.33
InChI Key IAPXDJMULQXGDD-NSHDSACASA-N
Synonym boc-asn-onp, boc-l-asparagine 4-nitrophenyl ester, boc-asn-ophno2, s-4-nitrophenyl 4-amino-2-tert-butoxycarbonyl amino-4-oxobutanoate, 4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-carbamoylpropanoate, pubchem12095, threonin-tert.-butylether, ch3 3coconhch cooh ch2cooh, n1-t-boc c1-p-nitrophenol asparagine, boc-asn-onp hplc
PubChem CID 439663
ChEBI CHEBI:3147
IUPAC Name (4-nitrophenyl) (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoate
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)OC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 136°C to 140°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Specific Rotation -36.4
Water 1% Max.
Quantity 1 g
Formula Weight 353.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-α-BOC-L-Asparagine 4-nitrophenyl ester
Show More Show Less
Product Identifier
  • N-alpha-BOC-L-Asparagine 4-nitrophenyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.