missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alanine methyl ester, 95%

Product Code. p-7054233
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448818 5 g 5g
15458818 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448818 Supplier Thermo Scientific Alfa Aesar Supplier No. H62271.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28875-17-4 | C9H17NO4 | 203.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28875-17-4
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00038513
InChI Key GJDICGOCZGRDFM-LURJTMIESA-N
Synonym boc-ala-ome, boc-l-alanine methyl ester, s-n-tert-butoxycarbonylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino propanoate, n-boc-l-alanine methyl ester, boc-alanine methyl ester, s-methyl 2-tert-butoxycarbonylamino propanoate, methyl n-tert-butoxycarbonyl-l-alaninate, methyl 2s-2-tert-butoxycarbonyl amino propanoate, methyl 2s-2-tert-butoxy carbonyl amino propanoate
PubChem CID 10856577
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}propanoate
SMILES COC(=O)[C@H](C)NC(=O)OC(C)(C)C

Specifications

Melting Point 32°C to 35°C
Color White
Beilstein 1872545
Density 1.030 g/mL
Quantity 5 g
Flash Point 113°C (235°F)
Formula Weight 203.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-L-alanine methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.