missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alanine methyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15458818
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28875-17-4
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00038513
InChI Key GJDICGOCZGRDFM-LURJTMIESA-N
Synonym boc-ala-ome, boc-l-alanine methyl ester, s-n-tert-butoxycarbonylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino propanoate, n-boc-l-alanine methyl ester, boc-alanine methyl ester, s-methyl 2-tert-butoxycarbonylamino propanoate, methyl n-tert-butoxycarbonyl-l-alaninate, methyl 2s-2-tert-butoxycarbonyl amino propanoate, methyl 2s-2-tert-butoxy carbonyl amino propanoate
PubChem CID 10856577
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}propanoate
SMILES COC(=O)[C@H](C)NC(=O)OC(C)(C)C

Specifications

Melting Point 32°C to 35°C
Color White
Beilstein 1872545
Formula Weight 203.24
Percent Purity 95%
Density 1.030 g/mL
Quantity 25 g
Flash Point 113°C (235°F)
Physical Form Powder
Chemical Name or Material N-Boc-L-alanine methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'