missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-guanidine, 95%, Thermo Scientific Chemicals

Product Code. 15449778
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in most alcohols, esters (e.g., ethyl acetate), ketones (e.g., acetone), acetonitrile, and dichloromethane. Not soluble in diethyl ether and hexane.

Notes
Moisture, heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219511-71-4
Molecular Formula C6H13N3O2
Molecular Weight (g/mol) 159.189
MDL Number MFCD20547465
InChI Key UMOZLQVSOVNSCA-UHFFFAOYSA-N
Synonym boc-guanidine, n-boc-guanidine, 1-tert-butoxycarbonyl guanidine, tert-butyl n-carbamimidoylcarbamate, boc guanidine, 1-boc-guanidine, tert-butyl n-diaminomethylidene carbamate, tert-butoxycarbonylguanidine, acmc-209x9z
PubChem CID 11378649
IUPAC Name tert-butyl N-(diaminomethylidene)carbamate
SMILES CC(C)(C)OC(=O)N=C(N)N

Specifications

Melting Point 170°C to 175°C
Color White
Linear Formula (CH3)3OCONHC(=NH)NH2
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in most alcohols,esters (e.g.,ethyl acetate),ketones (e.g.,acetone),acetonitrile,and dichloromethane. Not soluble in diethyl ether and hexane.
Formula Weight 159.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-guanidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'