missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals

Product Code. 15469338
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469338 5 g 5g
15479338 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469338 Supplier Thermo Scientific Alfa Aesar Supplier No. H62889.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31972-52-8
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00038558
InChI Key HWBAHOVOSOAFLE-UHFFFAOYSA-N
Synonym boc-gly-gly-oh, boc-gly-gly, n-tert-butoxycarbonyl glycylglycine, n-2-n-boc-amino-acetyl-glycine, boc-glycylglycine, 2-2-tert-butoxycarbonyl amino acetamido acetic acid, 2-tert-butoxycarbonyl amino acetamido acetic acid, 2-2-tert-butoxycarbonylamino acetamido acetic acid, 2-2-tert-butoxy carbonyl amino acetamido acetic acid, t-boc-glygly
PubChem CID 333466
IUPAC Name 2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)NCC(=O)NCC(=O)O

Specifications

Color White
Linear Formula (CH3)3COCONHCH2CONHCH2CO2H
Quantity 5 g
Formula Weight 232.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Boc-glycylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.