missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals

Product Code. 15479338
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31972-52-8
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00038558
InChI Key HWBAHOVOSOAFLE-UHFFFAOYSA-N
Synonym boc-gly-gly-oh, boc-gly-gly, n-tert-butoxycarbonyl glycylglycine, n-2-n-boc-amino-acetyl-glycine, boc-glycylglycine, 2-2-tert-butoxycarbonyl amino acetamido acetic acid, 2-tert-butoxycarbonyl amino acetamido acetic acid, 2-2-tert-butoxycarbonylamino acetamido acetic acid, 2-2-tert-butoxy carbonyl amino acetamido acetic acid, t-boc-glygly
PubChem CID 333466
IUPAC Name 2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)NCC(=O)NCC(=O)O

Specifications

Color White
Linear Formula (CH3)3COCONHCH2CONHCH2CO2H
Formula Weight 232.23
Percent Purity 97%
Quantity 25 g
Physical Form Powder
Chemical Name or Material N-Boc-glycylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'