missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Glycine methyl ester, 97%

Product Code. p-3770733 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10473074

Brand: Thermo Scientific Acros 399380050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 31954-27-5 | C8H15NO4 | 189.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31954-27-5
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00038267
InChI Key PHUZOEOLWIHIKH-UHFFFAOYSA-N
Synonym boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate
PubChem CID 637609
IUPAC Name methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CC(C)(C)OC(=O)NCC(=O)OC

Specifications

Boiling Point 190.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Glass Bottle
Density 1.2800g/mL
Quantity 5 mL
Flash Point 109°C
Formula Weight 189.21
Percent Purity 97%
Specific Gravity 1.28
Chemical Name or Material N-BOC-Glycine methyl ester
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.