missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine methyl ester, 97%

Product Code. 15485478
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485478 25 mL 25mL
15475478 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485478 Supplier Thermo Scientific Alfa Aesar Supplier No. H59234.AC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31954-27-5 | C8H15NO4 | 189.211 g/mol

It is used as a pharmaceutical, chemical, paint and dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical, chemical, paint and dye intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31954-27-5
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00038267
InChI Key PHUZOEOLWIHIKH-UHFFFAOYSA-N
Synonym boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate
PubChem CID 637609
IUPAC Name methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CC(C)(C)OC(=O)NCC(=O)OC

Specifications

Density 1.079
Boiling Point 190°C
Flash Point 109°C (228°F)
Linear Formula (CH3)3COCONHCH2CO2CH3
Refractive Index 1.437
Quantity 25 mL
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material N-Boc-glycine methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.