missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-gamma-aminobutyric acid, 98+%

Product Code. 11480527
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480527 5 g 5g
11470527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480527 Supplier Thermo Scientific Alfa Aesar Supplier No. B22407.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57294-38-9 | C9H17NO4 | 203.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57294-38-9
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00037313
InChI Key HIDJWBGOQFTDLU-UHFFFAOYSA-N
Synonym boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid
PubChem CID 294894
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NCCCC(O)=O

Specifications

Melting Point 57°C to 60°C
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98%
Chemical Name or Material N-Boc-gamma-aminobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.