missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-gamma-Aminobutyric acid, 98%, Thermo Scientific Chemicals

Catalog Number 10718333
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
57294-38-9
C9H17NO4
203.24
MFCD00037313
HIDJWBGOQFTDLU-UHFFFAOYSA-N
boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid
294894
4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)OC(=O)NCCCC(O)=O
This item is not returnable. View return policy
57294-38-9
C9H17NO4
203.24
MFCD00037313
HIDJWBGOQFTDLU-UHFFFAOYSA-N
boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid
294894
4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CC(C)(C)OC(=O)NCCCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 55°C
Boiling Point 78°C to 82°C (0.2mmHg)
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula (CH3)3COC(=O)NH(CH2)3CO2H
Packaging Glass Bottle
Formula Weight 203.24
Percent Purity 98%
Quantity 1 g
Flash Point >110°C
Physical Form Powder
Chemical Name or Material N-BOC-γ-Aminobutyric acid
Show More Show Less
Product Identifier
  • N-BOC-gamma-Aminobutyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'