missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-pipecolinic acid, 98%

Product Code. 11358486
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358486 5 g 5g
11348486 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358486 Supplier Thermo Scientific Alfa Aesar Supplier No. L17528.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98303-20-9 | C11H19NO4 | 229.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98303-20-9
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD01862877
InChI Key JQAOHGMPAAWWQO-UHFFFAOYSA-N
Synonym n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
PubChem CID 581831
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

Melting Point 129°C to 131°C
Quantity 5 g
Beilstein 480022
Formula Weight 229.28
Percent Purity 98%
Chemical Name or Material N-Boc-DL-pipecolinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.