missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-DL-2-Aminotetralin-2-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10026243 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10026243 5 g 5g
10143631 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10026243 Supplier Thermo Scientific Acros Supplier No. 334370050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 98569-12-1
MDL Number MFCD00800586
InChI Key DSDQZWPRZHNCIX-UHFFFAOYSA-N
Synonym 2-boc-amino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 2-n-boc-amino-tetrahydro-2-naphthoic acid, 2-tert-butoxycarbonylamino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, n-boc-dl-2-aminotetralin-2-carboxylic acid, 2-n-boc-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 2-tert-butoxycarbonyl amino-3,4-dihydro-1h-naphthalene-2-carboxylic acid, 2-tert-butoxy carbonylamino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 2-tert-butoxycarbonyl amino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, boc-atc
PubChem CID 2733177
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3,4-dihydro-1H-naphthalene-2-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCC2=CC=CC=C2C1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C16H21NO4
Quantity 5 g
Molecular Weight (g/mol) 291.347
Formula Weight 291.35
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-DL-2-Aminotetralin-2-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.