missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-D-Serine methyleester, 97%, Thermo Scientific Chemicals

Product Code. 10224574 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10224574 10 g Glass Bottle 10g
10402894 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10224574 Supplier Thermo Scientific Acros Supplier No. 399370100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95715-85-8
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00270516
InChI Key SANNKFASHWONFD-ZCFIWIBFSA-N
Synonym boc-d-ser-ome, boc-d-serine methyl ester, n-boc-d-serine methyl ester, n-boc-d-serine methylester, n-tert-butoxycarbonyl-d-serine methyl ester, boc-d-serinemethyl ester, n-boc-d-serinemethylester, --methyl n-boc-d-serinate, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl n-tert-butoxycarbonyl-d-serinate
PubChem CID 377723
IUPAC Name methyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC

Specifications

Boiling Point 215.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Glass Bottle
Density 1.0800g/mL
Quantity 10 g
Flash Point 113°C
Formula Weight 219.24
Percent Purity 97%
Specific Gravity 1.08
Chemical Name or Material N-BOC-D-Serine methyl ester
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-D-Serine methyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.