missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-prolinol, 99%, Thermo Scientific™

Product Code. 15469029
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469029 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15469029 Supplier Thermo Scientific Alfa Aesar Supplier No. L16268.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 83435-58-9
Melting Point 59°C to 62°C
Molecular Formula C10H19NO3
MDL Number MFCD00040580
Quantity 1 g
Synonym boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate
InChI Key BFFLLBPMZCIGRM-MRVPVSSYSA-N
Optical Rotation +54° (c=5 in Methanol)
SMILES CC(C)(C)OC(=O)N1CCCC1CO
IUPAC Name tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular Weight (g/mol) 201.266
PubChem CID 688279
Formula Weight 201.27
Percent Purity 99%
Chemical Name or Material N-Boc-D-prolinol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.